
2,6-Dichlorostyrene 96% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:28469-92-3
:
Formula:C₈H₆Cl₂
MDL Number:MFCD00000579
:173.036 g/mol
Boiling Pt:88…90 °C (8 mmHg)
Density:1.266
Flash Pt:71 °C (159 °F)
:Stabilised with 4-tert-butylpyrocatechol (TBC) 0.1%
EINECS:249-039-7
:GHS07
Storage Temperature:Refrigerator