

1,3-Dichloro-1,1,3,3-tetramethyldisiloxane 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2401-73-2
:
Formula:C₄H₁₂Cl₂OSi₂
MDL Number:MFCD00018088
:203.21 g/mol
Boiling Pt:136…138 °C
Melting Pt:–37 °C
Density:1.039
Flash Pt:15 °C (59 °F)
Storage Temperature:Ambient
EINECS:219-278-1
ADR:3,II
:GHS02
:GHS05