0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Berberine chloride hydrate
Berberine chloride hydrate, (∼17% H₂O) 96%
Berberine chloride hydrate, (∼17% H₂O) 96%
:
Thermo Fisher Scientific
Berberine chloride hydrate, (∼17% H₂O) 96%
:
Thermo Fisher Scientific
:
Natural Yellow 18
:
141433-60-5
:
:
C₂₀H₁₈ClNO₄
:
MFCD00011939
:
371.82 g/mol
:
14,1154
:
∼ 200 °C (dec.)
:
Ambient
:
211-195-9
Environmentally Preferable:
msds_ALFAL03807
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years