Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Gold
Gold 95% supplied in 0.1 mM PBS, nanoparticles 80 nm OD1 (Abs. 553 nm)
Gold 95% supplied in 0.1 mM PBS, nanoparticles 80 nm OD1 (Abs. 553 nm)
Gold 95% supplied in 0.1 mM PBS, nanoparticles 80 nm OD1 (Abs. 553 nm)
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Gold 95% supplied in 0.1 mM PBS, nanoparticles 80 nm OD1 (Abs. 553 nm)
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
7440-57-5
:
Formula:
Au
MDL Number:
MFCD00003436
:
196.97 g/mol
Storage Temperature:
Refrigerator
Related Information
Documentation
Certificates
Search by Lot Number
Videos
Thermo Scientific Acroseal 25 Years
Documentation