PYR-41
Supplier: Thermo Fisher Scientific
Jump to:
About this item
PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity.
- Light Sensitive
Dark purple Solid
Applications: PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity.
Soluble in DMSO at 5mg/ml with warming
Synonyms: Ethyl 4-(4-((5-nitrofuran-2-yl)methylene)-3,5-dioxopyrazolidin-1-yl)benzoate
- CAS Number:418805-02-4
- Formula:C₁₇H₁₃N₃O₇·3H₂O
- MDL Number:MFCD01469983
- Molecular Weight:425.40 g/mol
- Storage Temperature:Freezer
2 Options Available Below
Product Details & Documents
PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity.
- Light Sensitive
Dark purple Solid
Applications: PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity.
Soluble in DMSO at 5mg/ml with warming
Synonyms: Ethyl 4-(4-((5-nitrofuran-2-yl)methylene)-3,5-dioxopyrazolidin-1-yl)benzoate
- CAS Number:418805-02-4
- Formula:C₁₇H₁₃N₃O₇·3H₂O
- MDL Number:MFCD01469983
- Molecular Weight:425.40 g/mol
- Storage Temperature:Freezer



