5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyinosine 98%
Catalog # J65136.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:554.59 g/mol
Storage Temperature:Freezer
EINECS:298-195-2
CAS Number:93778-57-5
:MFCD00057879
Formula:C₃₁H₃₀N₄O₆
MDL Number:MFCD00080301