PJ-34 hydrochloride
Catalog # J64413.MB
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:295.2·36.5 g/mol
Storage Temperature:Freezer
CAS Number:344458-15-7
:MFCD16618379
Formula:C₁₇H₁₇N₃O₂·HCl
MDL Number:MFCD22581510