Cresyl violet (acetate)
Catalog # J64318.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 g
Specifications
About this item
:
:321.33 g/mol
Melting Pt:140…143 °C
Storage Temperature:Ambient
EINECS:234-043-3
CAS Number:10510-54-0
Formula:C₁₈H₁₅N₃O₃
MDL Number:MFCD00013151