5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-N2-isobutyryl-2'-deoxyguanosine 98%
Catalog # J64232.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:657.69 g/mol
Storage Temperature:Refrigerator
Formula:C₃₅H₃₆FN₅O₇
MDL Number:MFCD15145247