

Apramycin sulphate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65710-07-8
:
Formula:C₂₁H₄₁N₅O₁₁·H₂SO₄
MDL Number:MFCD06200257
Merck Index:14,752
Boiling Pt:823 °C
Melting Pt:168 °C
Density:1.56
Flash Pt:451.6 °C (845 °F)
:637.65 g/mol
EINECS:265-890-7
:GHS08
:GHS07