Yeast lytic enzyme
Applications: Hydrolyzes linear glucose polymers with beta-1,3-linkages
- CAS No.:37340-57-1
- Storage conditions:2...8 °C
- Form:White to off-White powder
- MDL No.:MFCD00131560
- Enzyme name:Yeast Lytic Enzyme
Applications: Hydrolyzes linear glucose polymers with beta-1,3-linkages