Order Entry
Export
ContactUsLinkComponent
Calcimycin (A23187) 98%
Calcimycin (A23187) 98%
Catalog # J63020.MA
CAS Number:  
Calcimycin (A23187) 98%
Catalog # J63020.MA
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :523.63 g/mol
Merck Index:14,1639
EINECS:258-084-1
 :GHS07
Storage Temperature:Refrigerator
CAS Number:52665-69-7
Formula:C₂₉H₃₇N₃O₆
MDL Number:MFCD00151202