L-thyroxine 98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 51-48-9 Molecular Formula: C15H11I4NO4 Formula Weight: 776.87 Physical Form: Crystalline powder Sensitivity: Light Sensitive Appearance: White to pale brown Odour: Odourless Melting Point: 235-236° Solubility: Dissolves in 4M ammonium hydroxide in Methanol at 50mg/ml Beilstein Reference: 2228515 MDL No.: MFCD00002595
Synonyms: (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
- CAS Number:51-48-9
- Formula:C₁₅H₁₁I₄NO₄
- MDL Number:MFCD00002595
- Molecular Weight:776.874 g/mol
- Merck Index:14,9415
- Melting Pt:235…236 °C
- Storage Temperature:Ambient
- EINECS:200-101-1
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 51-48-9 Molecular Formula: C15H11I4NO4 Formula Weight: 776.87 Physical Form: Crystalline powder Sensitivity: Light Sensitive Appearance: White to pale brown Odour: Odourless Melting Point: 235-236° Solubility: Dissolves in 4M ammonium hydroxide in Methanol at 50mg/ml Beilstein Reference: 2228515 MDL No.: MFCD00002595
Synonyms: (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
- CAS Number:51-48-9
- Formula:C₁₅H₁₁I₄NO₄
- MDL Number:MFCD00002595
- Molecular Weight:776.874 g/mol
- Merck Index:14,9415
- Melting Pt:235…236 °C
- Storage Temperature:Ambient
- EINECS:200-101-1



