7-Methoxycoumarin-4-acetic acid
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 7-Methoxy-2-oxo-2H-1-benzopyran-4-acetic acid, Mca, 2-(7-Methoxy-2-oxo-2H-chromen-4-yl)acetic acid, (7-Methoxycoumarin-4-yl)acetic acid
CAS Number:62935-72-2
:
Formula:C₁₂H₁₀O₅
MDL Number:MFCD00009774
:234.207 g/mol
Melting Pt:193 °C
Flash Pt:87 °C (189 °F)
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: