Histamine diphosphate 98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99+% CAS No.: 51-74-1 Molecular Formula: C5H9N3.2H3PO4 Formula Weight: 307.14 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 128-132° MDL No.: MFCD00064316
Applications: Endogenous H1 and H2 agonist
Synonyms: 2-(4-Imidazolyl)ethylamine diphosphate salt, 2-(1H-Imidazol-4-yl)ethan-1-amine bis(phosphate), Histamine phosphate
- CAS Number:51-74-1
- Formula:C₅H₉N₃·2H₃PO₄
- MDL Number:MFCD00064316
- Molecular Weight:307.14 g/mol
- Merck Index:14,4719
- Melting Pt:128…132 °C
- Storage Temperature:Refrigerator
- EINECS:200-118-4
2 Options Available Below
Product Details & Documents
Purity: 99+% CAS No.: 51-74-1 Molecular Formula: C5H9N3.2H3PO4 Formula Weight: 307.14 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 128-132° MDL No.: MFCD00064316
Applications: Endogenous H1 and H2 agonist
Synonyms: 2-(4-Imidazolyl)ethylamine diphosphate salt, 2-(1H-Imidazol-4-yl)ethan-1-amine bis(phosphate), Histamine phosphate
- CAS Number:51-74-1
- Formula:C₅H₉N₃·2H₃PO₄
- MDL Number:MFCD00064316
- Molecular Weight:307.14 g/mol
- Merck Index:14,4719
- Melting Pt:128…132 °C
- Storage Temperature:Refrigerator
- EINECS:200-118-4
Documents
Product Family Options
Product Information
- PkAvailabilityPrice



