Order Entry
Export
ContactUsLinkComponent
Phosphatase inhibitor cocktail II
Phosphatase inhibitor cocktail II
Catalog # J61022.AA
CAS Number:  
Phosphatase inhibitor cocktail II
Catalog # J61022.AA
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pk:
    5 mL
  • Description:
    Phosphatase inhibitor cocktail II
  • Storage temperature:
    -30°C to -10°C
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

About this item

The phosphatase inhibitor cocktail improves the yields of intact proteins by adding inhibitors to enzymes that modify proteins present in cell extracts.

  • Storage temperature: –30 to –10 °C
  • Physical form: Liquid
  • Appearance: Colourless

This product has been optimized and tested for tyrosine protein phosphatases, acid and alkaline phosphatases. It has been tested on cell extracts from animal tissues including bovine liver and human placenta, and cytosolic extract of rabbit muscle. It is a featured product for Kinase Phosphatase Biology research.An inhibitor of L-isozymes of alkaline phosphatase and ser/thr protein phosphatases. It is used with tissue and cell extracts, including extracts containing detergents.

Delivery: Contains 100 mM sodium fluoride, 50 mM sodium orthovanadate, 50 mM sodium pyrophosphate, and 100 mM b-glycerophosphate. 100X concentrate.
Caution: This cocktail will inhibit broad spectrum of serine/theorine and tyrosine phosphatases and is suitable for cell and tissue lysis. Store at –20 °C.