Order Entry
Export
Orders LinkContactUsLinkComponent
 

Miconazole 97%

Miconazole 97%
Certificates
Miconazole 97%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 22916-47-8 Molecular Formula: C18H14Cl4N2O Formula Weight: 416.13 Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Light Sensitive Appearance: White to off-white Melting Point: 170° Solubility: Freely soluble in alcohols or acetone. Also soluble in DMF or chloroform. Insoluble in water MDL No.: MFCD00216019

Applications: Inhibits cytochrome P450-dependent 14-alpha-demethylase

Synonyms:  1-(2-((2,4-Dichlorobenzyl)oxy)-2-(2,4-dichlorophenyl)ethyl)-1H-imidazole
  • CAS Number:22916-47-8
  • Formula:C₁₈H₁₄Cl₄N₂O
  • MDL Number:MFCD00216019
  • Molecular Weight:416.123 g/mol
  • Merck Index:14,6178
  • Melting Pt:83…97 °C
  • EINECS:245-324-5
Product Family Options

Product Information