Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Iminothiolane hydrochloride (Traut's Reagent)
2-Iminothiolane hydrochloride
2-Iminothiolane hydrochloride
2-Iminothiolane hydrochloride
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
2-Iminothiolane hydrochloride
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
4781-83-3
:
Formula:
C₄H₈ClNS
MDL Number:
MFCD00039013
:
137.625 g/mol
Melting Pt:
190…195 °C
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAJ60131
Videos
Thermo Scientific Acroseal 25 Years
Documentation