Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-4,5-difluorobenzaldehyde
2-Bromo-4,5-difluorobenzaldehyde 98%
2-Bromo-4,5-difluorobenzaldehyde 98%
Irritant
2-Bromo-4,5-difluorobenzaldehyde 98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
2-Bromo-4,5-difluorobenzaldehyde 98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
476620-54-9
:
Formula:
C₇H₃BrF₂O
MDL Number:
MFCD16659662
:
221.001 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation