
2,4-Dichloro-6-nitrophenol 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:609-89-2
:
Formula:C₆H₃Cl₂NO₃
MDL Number:MFCD00007101
:208 g/mol
Melting Pt:118…120 °C
Storage Temperature:Ambient
EINECS:210-202-2
ADR:6.1,III
:GHS07