Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-4,6-dichloronicotinate
Ethyl-4,6-dichloronicotinate 95%
Ethyl-4,6-dichloronicotinate 95%
H63092.06
:
Thermo Fisher Scientific
:
Ethyl-4,6-dichloronicotinate 95%
H63092.06
:
Thermo Fisher Scientific
:
Pk:
5 g
:
40296-46-6
:
:
C₈H₇Cl₂NO₂
:
220.05 g/mol
:
GHS07
:
MFCD00234408