Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dihydroxyanthraquinone-2-carboxylic acid
4,5-Dihydroxyanthraquinone-2-carboxylic acid 98%
4,5-Dihydroxyanthraquinone-2-carboxylic acid 98%
H60902.03
:
Thermo Fisher Scientific
:
4,5-Dihydroxyanthraquinone-2-carboxylic acid 98%
H60902.03
:
Thermo Fisher Scientific
:
Pk:
1 g
:
478-43-3
:
:
C₁₅H₈O₆
:
MFCD00009618
:
284.22 g/mol
:
>300 °C
:
207-521-4
:
GHS07