



3,4-Difluorobenzylzinc bromide 0.5 M in THF
Catalog # H58167.AD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
:272.398 g/mol
Density:0.98
ADR:4.3,II
:Packaged under argon in resealable ChemSeal™ bottles.
Storage Temperature:Refrigerator
UN:3399
:GHS02
:GHS05
:GHS08
:GHS07
CAS Number:307496-34-0
Formula:C₇H₅BrF₂Zn
MDL Number:MFCD01311420