Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Phenyl-1,4-diazepan-5-one
7-Phenyl-1,4-diazepan-5-one 97%
7-Phenyl-1,4-diazepan-5-one 97%
H57126.03
:
Thermo Fisher Scientific
:
7-Phenyl-1,4-diazepan-5-one 97%
H57126.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Seq:
3047
:
89044-79-1
:
:
C₁₁H₁₄N₂O
:
MFCD12197004
:
190.25 g/mol
:
104…106 °C
:
GHS07
:
Ambient