5,5'-Dibromo-2,2'-bithiophene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4805-22-5
:
Formula:C₈H₄Br₂S₂
MDL Number:MFCD00219110
:324.05 g/mol
Melting Pt:144…146 °C
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Organic Photovoltaics