Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-[(Trimethylsilyl)ethynyl]benzonitrile
3-[(Trimethylsilyl)ethynyl]benzonitrile 97%
3-[(Trimethylsilyl)ethynyl]benzonitrile 97%
:
Thermo Fisher Scientific
3-[(Trimethylsilyl)ethynyl]benzonitrile 97%
:
Thermo Fisher Scientific
:
190771-22-3
:
:
C₁₂H₁₃NSi
:
MFCD09260494
:
199.33 g/mol
:
42…47 °C
:
GHS07
:
Ambient
msds_ALFAH55012
Thermo Scientific Acroseal 25 Years