Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-5,6-dimethylpyrimidin-2-ylamine
4-Chloro-5,6-dimethylpyrimidin-2-ylamine 97%
4-Chloro-5,6-dimethylpyrimidin-2-ylamine 97%
Irritant
4-Chloro-5,6-dimethylpyrimidin-2-ylamine 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
4-Chloro-5,6-dimethylpyrimidin-2-ylamine 97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
14394-61-7
:
Formula:
C₆H₈ClN₃
MDL Number:
MFCD00462111
:
157.6 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH54552
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation