
2-Chlorodibenzo[b,f]-1,4-oxazepin-11(10H)-one 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3158-91-6
:
Formula:C₁₃H₈ClNO₂
MDL Number:MFCD06658244
:245.66 g/mol
EINECS:221-602-1
:GHS07
Storage Temperature:Ambient