Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,7-Dibromo-1-indanone
5,7-Dibromo-1-indanone 97%
5,7-Dibromo-1-indanone 97%
Irritant
5,7-Dibromo-1-indanone 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
5,7-Dibromo-1-indanone 97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
923977-18-8
:
Formula:
C₉H₆Br₂O
MDL Number:
MFCD11707100
:
289.95 g/mol
Melting Pt:
148 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation