Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Di(2-pyridyl)-1,2,4,5-tetrazine
3,6-Di(2-pyridyl)-1,2,4,5-tetrazine 96%
3,6-Di(2-pyridyl)-1,2,4,5-tetrazine 96%
:
Thermo Fisher Scientific
3,6-Di(2-pyridyl)-1,2,4,5-tetrazine 96%
:
Thermo Fisher Scientific
:
1671-87-0
:
:
C₁₂H₈N₆
:
MFCD00121717
:
236.24 g/mol
:
225 °C (dec.)
:
GHS07
:
Ambient
msds_ALFAH53484
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years