
3,4-Dimethoxy-L-phenylalanine 97%
Catalog # H53368.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:32161-30-1
:
Formula:C₁₁H₁₅NO₄
MDL Number:MFCD01321274
:225.24 g/mol
Melting Pt:254…257 °C
:GHS07
Storage Temperature:Ambient