
Octahydro-2H-pyrido[1,2-a]pyrazine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4430-75-5
:
Formula:C₈H₁₆N₂
MDL Number:MFCD01098257
:140.23 g/mol
Boiling Pt:68…72 °C (3 mmHg)
Storage Temperature:Ambient
ADR:8,II
:GHS05