1,6-Naphthyridin-2(1H)-one 97%
Catalog # H50121.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:23616-29-7
:
Formula:C₈H₆N₂O
MDL Number:MFCD07781179
:146.15 g/mol
Storage Temperature:Ambient