Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Dibromo-3,4,5,6-tetramethylbenzene
1,2-Dibromo-3,4,5,6-tetramethylbenzene 98%
1,2-Dibromo-3,4,5,6-tetramethylbenzene 98%
Irritant
1,2-Dibromo-3,4,5,6-tetramethylbenzene 98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
1,2-Dibromo-3,4,5,6-tetramethylbenzene 98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
36321-73-0
:
Formula:
C₁₀H₁₂Br₂
MDL Number:
MFCD00154987
:
292.03 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH33558
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation