Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Furo[3,2-b]pyridine
Furo[3,2-b]pyridine 98%
Furo[3,2-b]pyridine 98%
:
Thermo Fisher Scientific
Furo[3,2-b]pyridine 98%
:
Thermo Fisher Scientific
:
272-62-8
:
:
C₇H₅NO
:
MFCD11870744
:
119.12 g/mol
:
79…80 °C (12 mmHg)
:
GHS07
:
Ambient
msds_ALFAH32966
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years