0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene
2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene 97%
2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene 97%
Irritant
2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene 97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
118754-55-5
:
Formula:
C₇H₂BrCl₂F₃O
MDL Number:
MFCD03412210
:
309.90 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH32580
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation