1,1'-Diethyl-2,2'-carbocyanine bromide 95%
Catalog # H32253.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:2670-67-9
:
Formula:C₂₅H₂₅BrN₂
MDL Number:MFCD00011973
:433.39 g/mol
:GHS07
Storage Temperature:Ambient