Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Celatom® FW-14
Celatom® FW-14
Celatom® FW-14
Health
Celatom® FW-14
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Health
Celatom® FW-14
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
68855-54-9
:
MDL Number:
MFCD00132803
EINECS:
272-489-0
:
GHS08
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH31733
Videos
Thermo Scientific Acroseal 25 Years
Documentation