

5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2002-04-2
:
Formula:C₇H₆N₄S
MDL Number:MFCD00473692
:178.22 g/mol
Melting Pt:242…248 °C
:GHS06
:GHS07
Storage Temperature:Ambient