Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Diphenyl(o-tolyl)phosphine
Diphenyl(o-tolyl)phosphine
Diphenyl(o-tolyl)phosphine
Diphenyl(o-tolyl)phosphine
H27503.03
:
Thermo Fisher Scientific
:
Diphenyl(o-tolyl)phosphine
H27503.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Seq:
3047
:
:
276.32 g/mol
:
67…70 °C
:
227-675-6
:
GHS07
:
Ambient
:
5931-53-3
:
C₁₉H₁₇P
:
MFCD00008515