
3,4-Dihydro-2H-1,5-benzodioxepine-7-carboxylic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20825-89-2
:
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00276310
:194.18 g/mol
Melting Pt:146…148 °C
:GHS07
Storage Temperature:Ambient