
3,5-Dimethyladamantane-1-carboxylic acid 97%
Catalog # H27073.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:208.3 g/mol
Melting Pt:107…109 °C
:GHS07
Storage Temperature:Ambient
CAS Number:14670-94-1
Formula:C₁₃H₂₀O₂
MDL Number:MFCD00188067