Order Entry
Export
ContactUsLinkComponent
1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane
Irritant
1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane
Catalog # H26643.MD
CAS Number:  
Irritant
1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane
Catalog # H26643.MD
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

CAS Number:214078-93-0
 :
Formula:C₂₄H₃₆N₄
MDL Number:MFCD09263312
 :380.58 g/mol
Melting Pt:67…68 °C
 :GHS07
Storage Temperature:Ambient