
Diethyl-2,5-dibromoadipate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:869-10-3
:MFCD00276565
Formula:C₁₀H₁₆Br₂O₄
MDL Number:MFCD00075379
:360.04 g/mol
Melting Pt:64…67 °C
Storage Temperature:Ambient
ADR:8,II
:GHS05