1,4-Dioxaspiro[4.5]decan-8-ol ≥90%
Catalog # H26037.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
Specifications
About this item
CAS Number:22428-87-1
:
Formula:C₈H₁₄O₃
MDL Number:MFCD00067003
:158.19 g/mol
Storage Temperature:Ambient