Order Entry
Export
ContactUsLinkComponent
1,4-Bis(trimethylsilyl)tetrafluorobenzene 98%
Irritant
1,4-Bis(trimethylsilyl)tetrafluorobenzene 98%
Catalog # H26009.03
CAS Number:  
Irritant
1,4-Bis(trimethylsilyl)tetrafluorobenzene 98%
Catalog # H26009.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:16956-91-5
 :
Formula:C₁₂H₁₈F₄Si₂
MDL Number:MFCD00093109
 :294.44 g/mol
Melting Pt:53…55 °C
 :GHS07
Storage Temperature:Ambient