1,2:3,4-Di-O-isopropylidene-ɑ-D-galactopyranose 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4064-06-6
:
Formula:C₁₂H₂₀O₆
MDL Number:MFCD00063225
Density:1.143
:260.29 g/mol
Storage Temperature:Ambient
EINECS:223-771-7