
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1046-56-6
:
Formula:C₂₀H₁₄N₄
MDL Number:MFCD00006462
:310.36 g/mol
Melting Pt:189…192 °C
EINECS:213-878-7
:GHS07
Storage Temperature:Ambient