Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Bis(2-hydroxyethoxy)benzene
1,4-Bis(2-hydroxyethoxy)benzene 95%
1,4-Bis(2-hydroxyethoxy)benzene 95%
1,4-Bis(2-hydroxyethoxy)benzene 95%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
1,4-Bis(2-hydroxyethoxy)benzene 95%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
104-38-1
:
Formula:
C₁₀H₁₄O₄
MDL Number:
MFCD00002861
:
198.22 g/mol
Melting Pt:
100…102 °C
Storage Temperature:
Ambient
EINECS:
203-197-3
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB24037
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation