Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tolazoline hydrochloride
Tolazoline hydrochloride 99%
Tolazoline hydrochloride 99%
B21764.14
:
Thermo Fisher Scientific
:
Tolazoline hydrochloride 99%
B21764.14
:
Thermo Fisher Scientific
:
Pk:
25 g
:
59-97-2
:
:
C₁₀H₁₃ClN₂
:
MFCD00012693
:
196.678 g/mol
:
14,9506
:
174…176 °C
:
200-447-3
:
GHS07
:
Ambient